siRNA Details
Gene Symbol: Zfyve9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183300.2 | NP_899123.2 | 7 | 820 | ||
GenBank mRNA | DQ303413.1 | - | 7 | 820 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfyve9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183300.2 | NP_899123.2 | 7 | 820 | ||
GenBank mRNA | DQ303413.1 | - | 7 | 820 |