siRNA Details
| Gene Symbol: Nt5c1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085502.1 | NP_001078971.1 | 3 | 417 | ||
| GenBank mRNA | BC147308.1 | - | 3 | 466 | 9056382 | |
| BC147309.1 | - | 3 | 466 | 9056378 | ||
| BC147730.1 | - | 3 | 461 | 9056679 | ||
| BC147733.1 | - | 3 | 461 | 9056682 | ||

Nt5c1a
Genomic Map