siRNA Details
| Gene Symbol: Zfp362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081098.1 | NP_001074567.1 | 9 | 1825 | ||
| GenBank mRNA | BC039093.1 | - | 5 | 1201 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp362| Gene Symbol: Zfp362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081098.1 | NP_001074567.1 | 9 | 1825 | ||
| GenBank mRNA | BC039093.1 | - | 5 | 1201 | ||