siRNA Details
| Gene Symbol: Pnck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017275.1 | NP_058971.1 | 5 | 555 | ||
| XM_006229572.2 | XP_006229634.1 | 5 | 387 | |||
| XM_006229573.2 | XP_006229635.1 | 5 | 411 | |||
| XM_006229574.2 | XP_006229636.1 | 5 | 361 | |||
| XM_006229575.2 | XP_006229637.1 | 5 | 365 | |||
| XM_008773628.1 | XP_008771850.1 | 5 | 480 | |||
| XM_008773629.1 | XP_008771851.1 | 4 | 358 | |||
| GenBank mRNA | AB004267.1 | - | 5 | 555 | ||
| D86556.1 | - | 5 | 406 | |||

Pnck
Genomic Map