siRNA Details
| Gene Symbol: Aim1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162970.1 | NP_001156442.1 | 2 | 1527 | ||
| GenBank mRNA | AK145690.1 | - | 2 | 1531 | ||
| BC150738.1 | - | 1 | 1456 | 9087649 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aim1l| Gene Symbol: Aim1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162970.1 | NP_001156442.1 | 2 | 1527 | ||
| GenBank mRNA | AK145690.1 | - | 2 | 1531 | ||
| BC150738.1 | - | 1 | 1456 | 9087649 | ||