siRNA Details
| Gene Symbol: Slc30a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039677.2 | NP_001034766.1 | 8 | 2372 | ||
| GenBank mRNA | AK031425.1 | - | 8 | 2167 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc30a2| Gene Symbol: Slc30a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039677.2 | NP_001034766.1 | 8 | 2372 | ||
| GenBank mRNA | AK031425.1 | - | 8 | 2167 | ||