siRNA Details
| Gene Symbol: Sh2d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099631.1 | NP_001093101.1 | 4 | 329 | ||
| GenBank mRNA | AK032598.1 | - | 4 | 263 | ||
| BC036961.1 | - | 5 | 363 | |||
| DQ832275.1 | - | 4 | 329 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh2d5| Gene Symbol: Sh2d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099631.1 | NP_001093101.1 | 4 | 329 | ||
| GenBank mRNA | AK032598.1 | - | 4 | 263 | ||
| BC036961.1 | - | 5 | 363 | |||
| DQ832275.1 | - | 4 | 329 | |||