siRNA Details
| Gene Symbol: Gm436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085504.1 | NP_001078973.1 | 4 | 967 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm436| Gene Symbol: Gm436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085504.1 | NP_001078973.1 | 4 | 967 | ||