siRNA Details
| Gene Symbol: Vps13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276465.1 | NP_001263394.1 | 70 | 14629 | ||
| NM_001276502.1 | NP_001263431.1 | 69 | 14545 | |||
| GenBank mRNA | AK134463.1 | - | 1 | 845 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps13d| Gene Symbol: Vps13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276465.1 | NP_001263394.1 | 70 | 14629 | ||
| NM_001276502.1 | NP_001263431.1 | 69 | 14545 | |||
| GenBank mRNA | AK134463.1 | - | 1 | 845 | ||