siRNA Details
| Gene Symbol: Vps13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276465.1 | NP_001263394.1 | 19 | 2824 | ||
| NM_001276502.1 | NP_001263431.1 | 18 | 2737 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps13d| Gene Symbol: Vps13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276465.1 | NP_001263394.1 | 19 | 2824 | ||
| NM_001276502.1 | NP_001263431.1 | 18 | 2737 | |||