siRNA Details
| Gene Symbol: Gm572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085505.1 | NP_001078974.1 | 4 | 301 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm572| Gene Symbol: Gm572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085505.1 | NP_001078974.1 | 4 | 301 | ||