siRNA Details
| Gene Symbol: Fam193a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243123.1 | NP_001230052.1 | 19, 20 | 4651 | ||
| GenBank mRNA | AK134942.1 | - | 1, 2 | 316 | ||
| BC099925.1 | - | 16, 17 | 3688 | 30653351 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam193a| Gene Symbol: Fam193a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243123.1 | NP_001230052.1 | 19, 20 | 4651 | ||
| GenBank mRNA | AK134942.1 | - | 1, 2 | 316 | ||
| BC099925.1 | - | 16, 17 | 3688 | 30653351 | ||