siRNA Details
| Gene Symbol: Fam193a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243123.1 | NP_001230052.1 | 21 | 4799 | ||
| GenBank mRNA | AK163625.1 | - | 2 | 112 | ||
| BC099925.1 | - | 18 | 3836 | 30653351 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam193a| Gene Symbol: Fam193a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243123.1 | NP_001230052.1 | 21 | 4799 | ||
| GenBank mRNA | AK163625.1 | - | 2 | 112 | ||
| BC099925.1 | - | 18 | 3836 | 30653351 | ||