siRNA Details
| Gene Symbol: A830010M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007574.2 | NP_001007575.2 | 6 | 5999 | ||
| NM_001168557.1 | NP_001162029.1 | 6 | 4030 | |||
| GenBank mRNA | AK036000.1 | - | 1 | 1769 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A830010M20Rik| Gene Symbol: A830010M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007574.2 | NP_001007575.2 | 6 | 5999 | ||
| NM_001168557.1 | NP_001162029.1 | 6 | 4030 | |||
| GenBank mRNA | AK036000.1 | - | 1 | 1769 | ||