siRNA Details
| Gene Symbol: Rimbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081388.1 | NP_001074857.1 | 1 | 166 | ||
| GenBank mRNA | AK044685.1 | - | 1 | 285 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rimbp2| Gene Symbol: Rimbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081388.1 | NP_001074857.1 | 1 | 166 | ||
| GenBank mRNA | AK044685.1 | - | 1 | 285 | ||