siRNA Details
| Gene Symbol: Pdap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033313.3 | NP_001028485.1 | 6 | 620 | ||
| GenBank mRNA | AK147170.1 | - | 6 | 621 | ||
| AK161518.1 | - | 5 | 749 | |||
| AK169233.1 | - | 6 | 554 | |||
| BC007162.1 | - | 5 | 520 | |||
| BC055927.1 | - | 6 | 534 | |||
| BC106168.1 | - | 5 | 508 | |||
| BC114598.1 | - | 4 | 354 | |||
| BC139204.1 | - | 6 | 573 | 8862226 | ||
| BC139205.1 | - | 6 | 573 | 8862227 | ||

Pdap1
Genomic Map