siRNA Details
| Gene Symbol: Uspl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013378.2 | NP_001013396.2 | 10 | 2942 | ||
| NM_001115149.1 | NP_001108621.1 | 9 | 3442 | |||
| NM_001115150.1 | NP_001108622.1 | 8 | 3273 | |||
| NM_001115151.1 | NP_001108623.1 | 9 | 2773 | |||
| NM_001115153.1 | NP_001108625.1 | 10 | 3619 | |||
| NM_001286824.1 | NP_001273753.1 | 9 | 3400 | |||
| NM_001286825.1 | NP_001273754.1 | 9 | 3384 | |||
| GenBank mRNA | AK088787.1 | - | 8 | 3139 | ||
| AK145409.1 | - | 10 | 3517 | |||
| AK146820.1 | - | 9 | 3267 | |||
| AK161654.1 | - | 9 | 2621 | |||
| AK168702.1 | - | 9 | 3267 | |||
| AK168981.1 | - | 9 | 3295 | |||
| BC028849.1 | - | 10 | 2836 | |||
| BC032291.1 | - | 7 | 4912 | |||
| BC086664.1 | - | 10 | 2785 | |||

Uspl1
Genomic Map