siRNA Details
| Gene Symbol: Tmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081111.2 | NP_001074580.1 | 5 | 1750 | ||
| GenBank mRNA | AK144729.1 | - | 5 | 1751 | ||
| BC150931.1 | - | 5 | 1761 | 9087842 | ||
| CN456538.1 | - | 3 | 293 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmf1| Gene Symbol: Tmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081111.2 | NP_001074580.1 | 5 | 1750 | ||
| GenBank mRNA | AK144729.1 | - | 5 | 1751 | ||
| BC150931.1 | - | 5 | 1761 | 9087842 | ||
| CN456538.1 | - | 3 | 293 | |||