siRNA Details
Gene Symbol: Tmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081111.2 | NP_001074580.1 | 15 | 3016 | ||
GenBank mRNA | BC150931.1 | - | 15 | 3027 | 9087842 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081111.2 | NP_001074580.1 | 15 | 3016 | ||
GenBank mRNA | BC150931.1 | - | 15 | 3027 | 9087842 |