siRNA Details
Gene Symbol: Ankrd26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081112.1 | NP_001074581.1 | 24 | 3347 | ||
GenBank mRNA | BC008111.1 | - | 4 | 893 | ||
BC056364.1 | - | 7 | 1406 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ankrd26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081112.1 | NP_001074581.1 | 24 | 3347 | ||
GenBank mRNA | BC008111.1 | - | 4 | 893 | ||
BC056364.1 | - | 7 | 1406 |