siRNA Details
| Gene Symbol: BC035044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254946.1 | NP_001241875.1 | 3 | 2101 | ||
| GenBank mRNA | AK088258.1 | - | 3 | 1945 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC035044| Gene Symbol: BC035044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254946.1 | NP_001241875.1 | 3 | 2101 | ||
| GenBank mRNA | AK088258.1 | - | 3 | 1945 | ||