siRNA Details
Gene Symbol: Klhl42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081237.1 | NP_001074706.1 | 3 | 5824 | ||
GenBank mRNA | AK129335.1 | - | 3 | 4725 | ||
BC011326.1 | - | 1 | 3308 | |||
BC030345.1 | - | 1 | 3319 |
