siRNA Details
Gene Symbol: Klhl42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081237.1 | NP_001074706.1 | 3 | 3996 | ||
GenBank mRNA | AK129335.1 | - | 3 | 2911 | ||
BC011326.1 | - | 1 | 1494 | |||
BC030345.1 | - | 1 | 1505 | |||
BC147633.1 | - | 3 | 3757 | 9056623 | ||
BC147636.1 | - | 3 | 3756 | 9056626 | ||
BC151064.1 | - | 3 | 3761 | 9087977 | ||
BC151065.1 | - | 3 | 3758 | 9087978 | ||
BC151068.1 | - | 3 | 3755 | 9087981 | ||
BC151076.1 | - | 3 | 3758 | 9087989 |
