siRNA Details
| Gene Symbol: Caprin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181541.4 | NP_853519.2 | 9 | 1756 | ||
| GenBank mRNA | AK140744.1 | - | 9 | 1759 | ||
| BC025636.1 | - | 9 | 1712 | |||
| BC125373.1 | - | 9 | 1645 | 40129888 | ||
| BC125375.1 | - | 9 | 1645 | 40129890 | ||

Caprin2
Genomic Map