siRNA Details
| Gene Symbol: Zbtb45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024699.1 | NP_001019870.1 | 2 | 456 | ||
| GenBank mRNA | AK154877.1 | - | 2 | 532 | ||
| AK155880.1 | - | 2 | 513 | |||
| AK166382.1 | - | 525 | ||||
| BC094359.1 | - | 2 | 456 | 6828341 | ||

Zbtb45
Genomic Map