siRNA Details
| Gene Symbol: Phldb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102613.1 | NP_001096083.1 | 6 | 952 | ||
| GenBank mRNA | BC052555.1 | - | 1 | 256 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phldb3| Gene Symbol: Phldb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102613.1 | NP_001096083.1 | 6 | 952 | ||
| GenBank mRNA | BC052555.1 | - | 1 | 256 | ||