siRNA Details
| Gene Symbol: Prr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175022.2 | NP_778187.2 | 10, 11 | 5615 | ||
| GenBank mRNA | AK032894.1 | - | 7, 8 | 2103 | ||
| AK173130.1 | - | 7, 8 | 5054 | |||
| BC025846.1 | - | 2, 3 | 776 | |||
| BC058674.1 | - | 7, 8 | 4254 | |||
| BC075628.1 | - | 11, 12 | 4525 | |||

Prr12
Genomic Map