siRNA Details
| Gene Symbol: Adamts17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033877.4 | NP_001029049.2 | 23 | 4134 | ||
| GenBank mRNA | AK135801.1 | - | 1 | 466 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamts17| Gene Symbol: Adamts17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033877.4 | NP_001029049.2 | 23 | 4134 | ||
| GenBank mRNA | AK135801.1 | - | 1 | 466 | ||