siRNA Details
| Gene Symbol: D430042O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081022.1 | NP_001074491.1 | 28 | 6073 | ||
| GenBank mRNA | AK052492.1 | - | 15 | 4072 | ||
| AK122309.1 | - | 11 | 4576 | |||
| BC030902.1 | - | 13 | 3317 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D430042O09Rik| Gene Symbol: D430042O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081022.1 | NP_001074491.1 | 28 | 6073 | ||
| GenBank mRNA | AK052492.1 | - | 15 | 4072 | ||
| AK122309.1 | - | 11 | 4576 | |||
| BC030902.1 | - | 13 | 3317 | |||