siRNA Details
Gene Symbol: Whsc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK048491.1 | - | 2 | 1552 | ||
AK079952.1 | - | 7 | 1328 | |||
AK156746.1 | - | 10 | 2567 | |||
BC064447.1 | - | 10 | 2605 |

More Information
Other Species Targeted

