siRNA Details
| Gene Symbol: Gucy1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033322.2 | NP_001028494.1 | 8 | 10144 | ||
| GenBank mRNA | AK038764.1 | - | 1 | 1464 | ||
| AK083446.1 | - | 1 | 220 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gucy1a2| Gene Symbol: Gucy1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033322.2 | NP_001028494.1 | 8 | 10144 | ||
| GenBank mRNA | AK038764.1 | - | 1 | 1464 | ||
| AK083446.1 | - | 1 | 220 | |||