siRNA Details
| Gene Symbol: Cep126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045524.1 | NP_001038989.1 | 6 | 1831 | ||
| GenBank mRNA | AK129341.1 | - | 4 | 1452 | ||
| BC119072.1 | - | 6 | 1831 | 8733821 | ||
| BC120489.1 | - | 6 | 1831 | 40129412 | ||

Cep126
Genomic Map