siRNA Details
| Gene Symbol: Tmem136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034863.3 | NP_001030035.1 | 3 | 3197 | ||
| GenBank mRNA | AK158698.1 | - | 3 | 3196 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem136| Gene Symbol: Tmem136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034863.3 | NP_001030035.1 | 3 | 3197 | ||
| GenBank mRNA | AK158698.1 | - | 3 | 3196 | ||