siRNA Details
| Gene Symbol: Zbtb16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033324.2 | NP_001028496.1 | 7 | 3656 | ||
| GenBank mRNA | AK142947.1 | - | 7 | 3657 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb16| Gene Symbol: Zbtb16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033324.2 | NP_001028496.1 | 7 | 3656 | ||
| GenBank mRNA | AK142947.1 | - | 7 | 3657 | ||