siRNA Details
| Gene Symbol: S1pr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017301.2 | NP_058997.1 | 2 | 520 | ||
| XM_008761459.1 | XP_008759681.1 | 2 | 572 | |||
| GenBank mRNA | BC097938.1 | - | 2 | 520 | 7460011 | |
| U10303.1 | - | 556 | ||||
More Information
Set Membership: |
|

S1pr1
Genomic Map