siRNA Details
Gene Symbol: Adgrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK144045.1 | - | 3, 4 | 645 | ||
BC132476.1 | - | 3, 4 | 493 | 40130753 | ||
BC132480.1 | - | 3, 4 | 493 | 40130757 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adgrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK144045.1 | - | 3, 4 | 645 | ||
BC132476.1 | - | 3, 4 | 493 | 40130753 | ||
BC132480.1 | - | 3, 4 | 493 | 40130757 |