siRNA Details
| Gene Symbol: Gclm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017305.2 | NP_059001.1 | 2, 3 | 321 | ||
| GenBank mRNA | BC078867.1 | - | 2, 3 | 321 | 7110139 | |
| DQ266366.1 | - | 1, 2 | 63 | |||
| L22191.1 | - | 2, 3 | 259 | |||
| S65555.1 | - | 2, 3 | 259 | |||

Gclm
Genomic Map