siRNA Details
Gene Symbol: Gm4925 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037166.2 | NP_001032243.1 | 1 | 1532 | ||
GenBank mRNA | BC100581.1 | - | 1 | 1564 | 6435651 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm4925 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037166.2 | NP_001032243.1 | 1 | 1532 | ||
GenBank mRNA | BC100581.1 | - | 1 | 1564 | 6435651 |