siRNA Details
| Gene Symbol: Gas2l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033331.2 | NP_001028503.2 | 11 | 2837 | ||
| NM_001079876.1 | NP_001073345.1 | 11 | 2840 | |||
| NM_001284344.1 | NP_001271273.1 | 12 | 1852 | |||
| GenBank mRNA | AK136054.1 | - | 11 | 2851 | ||
| AK136819.1 | - | 1 | 183 | |||
| AK164505.1 | - | 12 | 1841 | |||
| AK170634.1 | - | 11 | 2772 | |||
| AK170750.1 | - | 11 | 2762 | |||
| AK171905.1 | - | 1 | 54 | |||

Gas2l3
Genomic Map