siRNA Details
| Gene Symbol: Gas2l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033331.2 | NP_001028503.2 | 11 | 6582 | ||
| NM_001079876.1 | NP_001073345.1 | 11 | 6585 | |||
| NM_001284344.1 | NP_001271273.1 | 12 | 5597 | |||
| GenBank mRNA | AK018456.1 | - | 1 | 1135 | ||
| AK171905.1 | - | 2 | 3579 | |||

Gas2l3
Genomic Map