siRNA Details
| Gene Symbol: Zfp867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178417.3 | NP_848504.2 | 2 | 242 | ||
| GenBank mRNA | AK132062.1 | - | 2 | 243 | ||
| BC050078.1 | - | 2 | 221 | 30008012 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp867| Gene Symbol: Zfp867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178417.3 | NP_848504.2 | 2 | 242 | ||
| GenBank mRNA | AK132062.1 | - | 2 | 243 | ||
| BC050078.1 | - | 2 | 221 | 30008012 | ||