siRNA Details
| Gene Symbol: Sema3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017310.1 | NP_059006.1 | 3 | 304 | ||
| XM_006235989.2 | XP_006236051.1 | 5 | 691 | |||
| XM_006235990.2 | XP_006236052.1 | 4 | 544 | |||
| XM_008762702.1 | XP_008760924.1 | 5 | 553 | |||
| GenBank mRNA | X95286.1 | - | 3 | 304 | ||

Sema3a
Genomic Map