siRNA Details
| Gene Symbol: 1700071K01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033765.2 | NP_001028937.1 | 1 | 978 | ||
| GenBank mRNA | AK131752.1 | - | 1 | 979 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700071K01Rik| Gene Symbol: 1700071K01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033765.2 | NP_001028937.1 | 1 | 978 | ||
| GenBank mRNA | AK131752.1 | - | 1 | 979 | ||