siRNA Details
Gene Symbol: Slfn14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166028.1 | NP_001159500.1 | 4 | 1441 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slfn14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166028.1 | NP_001159500.1 | 4 | 1441 |