siRNA Details
| Gene Symbol: Gpatch8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159492.1 | NP_001152964.1 | 8 | 6302 | ||
| GenBank mRNA | AK017270.1 | - | 1 | 1373 | ||
| AK122307.1 | - | 1 | 3752 | |||
| AK142570.1 | - | 1 | 3547 | |||
| BC019948.1 | - | 1 | 813 | |||

Gpatch8
Genomic Map