siRNA Details
| Gene Symbol: Gpatch8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159492.1 | NP_001152964.1 | 8 | 969 | ||
| GenBank mRNA | BC016116.1 | - | 1 | 110 | ||
| BC035255.1 | - | 6 | 1028 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpatch8| Gene Symbol: Gpatch8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159492.1 | NP_001152964.1 | 8 | 969 | ||
| GenBank mRNA | BC016116.1 | - | 1 | 110 | ||
| BC035255.1 | - | 6 | 1028 | |||