siRNA Details
| Gene Symbol: Cog5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163126.1 | NP_001156598.1 | 6 | 467 | ||
| GenBank mRNA | AK030656.1 | - | 6 | 468 | ||
| CA316530.1 | - | 3 | 122 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cog5| Gene Symbol: Cog5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163126.1 | NP_001156598.1 | 6 | 467 | ||
| GenBank mRNA | AK030656.1 | - | 6 | 468 | ||
| CA316530.1 | - | 3 | 122 | |||