siRNA Details
| Gene Symbol: Rab3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017313.3 | NP_059009.2 | 1 | 257 | ||
| XM_006241357.2 | XP_006241419.1 | 2 | 426 | |||
| XM_006241358.2 | XP_006241420.1 | 2 | 431 | |||
| XM_006241359.2 | XP_006241421.1 | 2 | 1435 | |||
| XM_006241360.2 | XP_006241422.1 | 2 | 421 | |||
| XM_008765382.1 | XP_008763604.1 | 1 | 483 | |||
| GenBank mRNA | BC127500.1 | - | 1 | 260 | 7457420 | |
| U19181.1 | - | 1 | 250 | |||

Rab3ip
Genomic Map