siRNA Details
Gene Symbol: Elmsan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163501.1 | NP_001156973.1 | 13 | 5652 | ||
NM_001163502.1 | NP_001156974.1 | 13 | 6062 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Elmsan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163501.1 | NP_001156973.1 | 13 | 5652 | ||
NM_001163502.1 | NP_001156974.1 | 13 | 6062 |