siRNA Details
| Gene Symbol: Elmsan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163501.1 | NP_001156973.1 | 13 | 5499 | ||
| NM_001163502.1 | NP_001156974.1 | 13 | 5909 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Elmsan1| Gene Symbol: Elmsan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163501.1 | NP_001156973.1 | 13 | 5499 | ||
| NM_001163502.1 | NP_001156974.1 | 13 | 5909 | |||